36148
Mecoprop methyl ester
PESTANAL®, analytical standard
Synonym(s):
Methyl 2-(4-chloro-2-methylphenoxy)propioate
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.52
suitability
passes test for identity (NMR)
application(s)
agriculture
environmental
format
neat
SMILES string
COC(=O)C(C)Oc1ccc(Cl)cc1C
InChI
1S/C11H13ClO3/c1-7-6-9(12)4-5-10(7)15-8(2)11(13)14-3/h4-6,8H,1-3H3
InChI key
YWGAULPFWIQKRB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
application
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Skin Sens. 1
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
>212.0 °F
flash_point_c
> 100 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service