Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36130

Sigma-Aldrich

2,4-Dichlorophenol

purum, ≥97.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OH
CAS Number:
Molecular Weight:
163.00
Beilstein/REAXYS Number:
742467
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥97.0% (HPLC)

form

solid

bp

209-210 °C (lit.)

mp

39-43 °C
42-43 °C (lit.)

SMILES string

Oc1ccc(Cl)cc1Cl

InChI

1S/C6H4Cl2O/c7-4-1-2-6(9)5(8)3-4/h1-3,9H

InChI key

HFZWRUODUSTPEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

231.1 °F - closed cup

flash_point_c

110.6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service