Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36103

Supelco

Acetyleugenol

reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Eugenyl acetate, 4-Allyl-2-methoxyphenyl acetate, O-Acetyleugenol, Eugenol acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O3
CAS Number:
Molecular Weight:
206.24
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

reference material

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

refractive index

n20/D 1.518 (lit.)

bp

281-286 °C (lit.)

density

1.079 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

COc1cc(CC=C)ccc1OC(C)=O

InChI

1S/C12H14O3/c1-4-5-10-6-7-11(15-9(2)13)12(8-10)14-3/h4,6-8H,1,5H2,2-3H3

InChI key

SCCDQYPEOIRVGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

Acetyleugenol is a phytochemical compound belonging to the class of alkylbenzenes, a group of volatile plant toxins. It is widely found in consumed spices, aromatic herbs, and other plants including anise, basil, cinnamon, fennel, nutmeg, clove Syzygium aromaticum (L.) essential oil, pimento, rose, ylang-ylang, etc.[1][2]

Application

Acetyleugenol may be used as a reference material in pepper and its other varieties,[1] Longhu Rendan pills[3] and clove extract[4] using chromatography techniques.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

287.6 °F - closed cup

flash_point_c

142 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new strategy based on gas chromatography--high resolution mass spectrometry (GC--HRMS-Q-Orbitrap) for the determination of alkenylbenzenes in pepper
Rivera-Perez A, et al.
Food Chemistry, 126727-126727 (2020)
Acetyleugenol from Acacia nilotica (L.) exhibits a strong antibacterial activity and its phenyl and indole analogs show a promising anti-TB potential
Abdulhamid A, et al.
Microbiological research, 12, 1-15 (2020)
Gas chromatographic determination of eugenol in ethanol extract of cloves
Myint S, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 679(1-2), 193-195 (1996)
J Chromatogr B Analyt Technol Biomed Life Sci
Wang, et al.
Analytical Methods : Advancing Methods and Applications, 6(11), 3713-3719 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service