Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

360686

Sigma-Aldrich

2-(Phenylsulfonyl)ethanol

new

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2CH2CH2OH
CAS Number:
Molecular Weight:
186.23
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

assay

97%

form

liquid

refractive index

n20/D 1.555 (lit.)

bp

177 °C/2 mmHg (lit.)

density

1.557 g/mL at 25 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OCCS(=O)(=O)c1ccccc1

InChI

1S/C8H10O3S/c9-6-7-12(10,11)8-4-2-1-3-5-8/h1-5,9H,6-7H2

InChI key

PQVYYVANSPZIKE-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service