Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35570

Sigma-Aldrich

1,4-Dichlorobutane

puriss., ≥99.0% (GC)

Synonym(s):

Tetramethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)4Cl
CAS Number:
Molecular Weight:
127.01
Beilstein/REAXYS Number:
635673
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

puriss.

assay

≥99.0% (GC)

refractive index

n20/D 1.454 (lit.)
n20/D 1.454

bp

161-163 °C (lit.)

mp

−38 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

SMILES string

ClCCCCCl

InChI

1S/C4H8Cl2/c5-3-1-2-4-6/h1-4H2

InChI key

KJDRSWPQXHESDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

104.0 °F - closed cup

flash_point_c

40 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service