Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35488

Supelco

4,4′-DDM

PESTANAL®, analytical standard

Synonym(s):

Bis(4-chlorophenyl)methane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10Cl2
CAS Number:
Molecular Weight:
237.12
Beilstein/REAXYS Number:
1873121
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(Cc2ccc(Cl)cc2)cc1

InChI

1S/C13H10Cl2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11/h1-8H,9H2

InChI key

LQGSWLJZAKVBJH-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fredrick Orori Kengara et al.
Environmental science and pollution research international, 26(9), 8779-8788 (2019-02-04)
DDT transformation to DDD in soil is the most commonly reported pathway under anaerobic conditions. A few instances of DDT conversion to products other than DDD/DDE have been reported under aerobic conditions and hardly any under anaerobic conditions. In particular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service