Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35484

Supelco

4,4′-DDA

PESTANAL®, analytical standard

Synonym(s):

2,2-Bis(4-chlorophenyl)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10Cl2O2
CAS Number:
Molecular Weight:
281.13
Beilstein/REAXYS Number:
1913593
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)C(c1ccc(Cl)cc1)c2ccc(Cl)cc2

InChI

1S/C14H10Cl2O2/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8,13H,(H,17,18)

InChI key

YIOCIFXUGBYCJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kujawa et al.
Die Nahrung, 31(3), 253-257 (1987-01-01)
DDE is hydrogenated in the rat organism. The distribution of the compound is described. DDMU is degradated to DBP as well as transformed to DDA and DDOH. Excretion and distribution are studied.
M A Dawson et al.
The Journal of pharmacology and experimental therapeutics, 266(2), 673-677 (1993-08-01)
We assessed the interaction of several pesticides with renal organic anion transport based on inhibition of the active transepithelial transport of p-aminohippuric acid ([3H]PAH) by primary cultures of winter flounder proximal tubules. Four structurally similar chlorophenoxy acid herbicides were tested.
L A Hilgers et al.
International archives of allergy and applied immunology, 80(3), 320-325 (1986-01-01)
Modulation of delayed-type hypersensitivity reaction (DTH) in mice by synthetic adjuvants and the mode of their action were investigated. Intracutaneous injection of azobenzenearsonate coupled to phosphatidylethanolamine (A-PE) without adjuvant did not induce DTH. Administration of A-PE with the quaternary amines
Zhenshan Chen et al.
Marine pollution bulletin, 64(7), 1300-1308 (2012-06-12)
DDT, DDE and some additional lipophilic derivatives are recognized contaminants in sediments of Southern California Bight (SCB). Only about 10% of total DDTs discharged into the SCB are accounted for using available monitoring data (sediment, water, and biota). DDA represented
J Guilford et al.
Drug and chemical toxicology, 9(3-4), 239-252 (1986-01-01)
The possibility that lead could affect the metabolism of the insecticide 1,1-dichloro-2, 2-bis(p-chlorophenyl)ethane, DDD, was examined by studies of the effects of chronic oral Pb treatment on DDD conversion to 2,2-bis(p-chlorophenyl)acetic acid, DDA. Rats were given either distilled deionized water

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service