Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35407

Supelco

β-Zearalanol solution

~10 μg/mL in acetonitrile, analytical standard

Synonym(s):

(3S,7S)-3,4,5,6,7,8,9,10,11,12-Decahydro-7,14,16-trihydroxy-3-methyl-1H-2-benzoxacyclotetradecin-1-one solution, 2,4-Dihydroxy-6-(6β,10-dihydroxyundecyl)benzoic acid μ-lactone solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O5
CAS Number:
Molecular Weight:
322.40
Beilstein/REAXYS Number:
4844761
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

availability

not available in Canada

concentration

~10 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

C[C@H]1CCC[C@@H](O)CCCCCc2cc(O)cc(O)c2C(=O)O1

InChI

1S/C18H26O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h10-12,14,19-21H,2-9H2,1H3/t12-,14-/m0/s1

InChI key

DWTTZBARDOXEAM-JSGCOSHPSA-N

Application

β-Zearalanol may be used as an analytical reference standard for the determination of the analyte in traditional medicinal herbs,[1][2] human and animal urine[1] by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

purity : ≥96.0% (HPLC)

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B H Migdalof et al.
Xenobiotica; the fate of foreign compounds in biological systems, 13(4), 209-221 (1983-04-01)
The disposition of [3H]zeranol has been studied in the female Wistar rat, New Zealand rabbit, beagle dog, rhesus monkey and man. The blood elimination half-life of total radioactivity in rabbit was 26 h, monkey 18 h and man 22 h.
Erika Pfeiffer et al.
Molecular nutrition & food research, 54(10), 1468-1476 (2010-04-17)
Glucuronidation constitutes an important pathway in the phase II metabolism of the mycotoxin zearalenone (ZEN) and the growth promotor α-zearalanol (α-ZAL, zeranol), but the enzymology of their formation is yet unknown. In the present study, ZEN, α-ZAL and four of
Alberto Sánchez Arribas et al.
Electrophoresis, 30(3), 499-506 (2009-01-22)
A simple analytical scheme for the screening and quantification of zearalenone and its metabolites, alpha-zearalenol and beta-zearalenol, is reported. Extracts from maize flour samples were collected by supercritical fluid extraction and afterwards, they were analyzed by CE with amperometric detection.
H Malekinejad et al.
Veterinary research communications, 30(4), 445-453 (2006-02-28)
The enzymes 3alpha- and 3beta-hydroxysteroid dehydrogenase (3alpha- and 3beta-HSD) play a pivotal role in synthesis of various steroid hormones including oestradiol and testosterone. The structure of the mycotoxin zearalenone resembles many characteristics of steroids and binds to oestrogen receptors as
F M Launay et al.
Food additives and contaminants, 21(1), 52-62 (2004-01-28)
A method is described for the quantitative determination of the veterinary drug zeranol, its epimer taleranol and the mycotoxins zearalenone, alpha-zearalenol and beta-zearalenol in bovine urine. The method is based on liquid chromatography coupled to negative-ion electrospray mass spectrometry-mass spectrometry

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service