35401
Clenisopenterol hydrochloride
VETRANAL®, analytical standard
Synonym(s):
1-(4-Amino-3,5-dichlorophenyl)-2-(3-methylbutylamino)ethanol
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
forensics and toxicology
pharmaceutical (small molecule)
format
neat
SMILES string
Cl.CC(C)CCNCC(O)c1cc(Cl)c(N)c(Cl)c1
InChI
1S/C13H20Cl2N2O.ClH/c1-8(2)3-4-17-7-12(18)9-5-10(14)13(16)11(15)6-9;/h5-6,8,12,17-18H,3-4,7,16H2,1-2H3;1H
InChI key
HEVWUZFPJCXIOO-UHFFFAOYSA-N
Application
Recommended products
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service