35386
Sulfisoxazole-(phenyl-13C6)
VETRANAL®, analytical standard
Synonym(s):
4-Amino-N-(3,4-dimethyl-5-isoxazolyl)benzene-13C6-sulfonamide
About This Item
grade
analytical standard
Quality Level
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
forensics and toxicology
pharmaceutical (small molecule)
format
neat
mass shift
M+6
SMILES string
Cc1noc(NS(=O)(=O)[13c]2[13cH][13cH][13c](N)[13cH][13cH]2)c1C
InChI
1S/C11H13N3O3S/c1-7-8(2)13-17-11(7)14-18(15,16)10-5-3-9(12)4-6-10/h3-6,14H,12H2,1-2H3/i3+1,4+1,5+1,6+1,9+1,10+1
InChI key
NHUHCSRWZMLRLA-MSQIWANRSA-N
Application
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service