Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

35353

Supelco

Clorsulon-(phenyl13C6)

VETRANAL®, analytical standard

Synonym(s):

4-Amino-6-trichloroethenyl-1,3-benzene-13C6-disulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C2H8Cl3N3O4S2
CAS Number:
Molecular Weight:
386.61
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

mass shift

M+6

SMILES string

N[13c]1[13cH][13c](\C(Cl)=C(\Cl)Cl)[13c]([13cH][13c]1S(N)(=O)=O)S(N)(=O)=O

InChI

1S/C8H8Cl3N3O4S2/c9-7(8(10)11)3-1-4(12)6(20(14,17)18)2-5(3)19(13,15)16/h1-2H,12H2,(H2,13,15,16)(H2,14,17,18)/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

QOVTVIYTBRHADL-IDEBNGHGSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service