Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35349

Supelco

Iminoctadine albesilate

PESTANAL®, analytical standard

Synonym(s):

N,N‴-(Iminodi-8,1-octanediyl)bisguanidine tris(dodecylbenzenesulfonate), Iminoctadine tris(albesilate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H41N7 · 3C18H30O3S
CAS Number:
Molecular Weight:
1335.05
UNSPSC Code:
41116107

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service