35342
2,5-Dimethyl-celecoxib
VETRANAL®, analytical standard
Synonym(s):
2,5-Dimethylcelecoxib, 4-[5-(2,5-dimethylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]-benzenesulfonamide
About This Item
grade
analytical standard
Quality Level
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
forensics and toxicology
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
Cc1ccc(C)c(c1)-c2cc(nn2-c3ccc(cc3)S(N)(=O)=O)C(F)(F)F
InChI
1S/C18H16F3N3O2S/c1-11-3-4-12(2)15(9-11)16-10-17(18(19,20)21)23-24(16)13-5-7-14(8-6-13)27(22,25)26/h3-10H,1-2H3,(H2,22,25,26)
InChI key
NTFOSUUWGCDXEF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service