Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34791BC

Millipore

GDS Positive Control for Cronobacter Tq

BioControl®, For use with GDS Cronobacter II

Synonym(s):

Positive Control for PCR

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$74.90

$74.90

List Price$107.00Save 30%
Web-Only Promotion

Available to ship onMay 02, 2025Details


Request more information

Select a Size

Change View
5 ML
$74.90

About This Item

UNSPSC Code:
41122409
NACRES:
NA.85

$74.90

List Price$107.00Save 30%
Web-Only Promotion

Available to ship onMay 02, 2025Details


Request more information

Quality Level

form

liquid

usage

sufficient for 166 tests

shelf life

12 mo.

packaging

vial of 5 mL

manufacturer/tradename

BioControl Systems

storage condition

(Tightly closed)

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with BioControl® Systems GDS

storage temp.

2-8°C

suitability

Cronobacter spp.

General description

GDS Positive Control for Cronobacter Tq is used with the GDS Cronobacter Tq II assay, which is an automated nucleic acid amplification system for the detection of Cronobacter spp. in infant nutritional formula, ingredients, and environmental samples.

Application

GDS Positive Control for Cronobacter Tq is used for confirmation of Cronobacter spp. detection in Assurance® GDS assays.

Components

One vial, 5 mL, sufficient for 166 tests

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
BioControl is a registered trademark of BioControl Systems, Inc.

related product

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service