Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

347663

Sigma-Aldrich

Sodium nitrate

beads, 1-2 mm, 98%

Synonym(s):

Chile salpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaNO3
CAS Number:
Molecular Weight:
84.99
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

98%

form

beads

particle size

1-2 mm

mp

306 °C (dec.) (lit.)

solubility

water: soluble 874 g/L at 20 °C

SMILES string

[Na+].[O-][N+]([O-])=O

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

pictograms

Flame over circleExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Polyol synthesis of platinum nanoparticles: control of morphology with sodium nitrate.
Herricks T, et al.
Nano Letters, 4(12), 2367-2371 (2004)
E T Urbansky et al.
Environmental pollution (Barking, Essex : 1987), 112(3), 299-302 (2001-04-09)
Paleogeochemical deposits in northern Chile are a rich source of naturally occurring sodium nitrate (Chile saltpeter). These ores are mined to isolate NaNO3 (16-0-0) for use as fertilizer. Coincidentally, these very same deposits are a natural source of perchlorate anion
Calcined sodium nitrate/natural phosphate: an extremely active catalyst for the easy synthesis of chalcones in heterogeneous media.
Sebti S, et al.
Tetrahedron Letters, 42(45), 7953-7955 (2001)
Suna Choi et al.
Journal of controlled release : official journal of the Controlled Release Society, 131(1), 70-76 (2008-08-12)
One of the most important requirements for non-viral gene delivery systems is the ability to mediate high levels of gene expression with low toxicity. After the DNA/vector complexes are taken up by cells through endocytosis, DNA is typically contained within
Daryoush Afzali et al.
Journal of chromatography. A, 1247, 35-41 (2012-06-08)
In the present study, a new approach which uses immunoaffinity column clean-up combined with dispersive liquid-liquid microextraction (DLLME) is proposed for the preconcentration of ultra trace amounts of aflatoxins (B₁, B₂, G₁ and G₂). The aflatoxins are then determined using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service