Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34684

Sigma-Aldrich

Dibutyldimethoxytin

technical, ≥90% Sn basis

Synonym(s):

Dibutyltin dimethoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]2Sn(OCH3)2
CAS Number:
Molecular Weight:
295.01
Beilstein/REAXYS Number:
4129589
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

technical

assay

≥90% Sn basis

reaction suitability

core: tin

refractive index

n20/D 1.487 (lit.)
n20/D 1.487

density

1.286 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(OC)OC

InChI

1S/2C4H9.2CH3O.Sn/c2*1-3-4-2;2*1-2;/h2*1,3-4H2,2H3;2*1H3;/q;;2*-1;+2

InChI key

ZXDVQYBUEVYUCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service