34624
Mercury(II) sulfate-Sulfuric acid solution
200 g/L
About This Item
Recommended Products
Quality Level
form
liquid
concentration
195-205 g/L
application(s)
environmental
general analytical
SMILES string
[Hg++].[O-]S([O-])(=O)=O
InChI
1S/Hg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2
InChI key
DOBUSJIVSSJEDA-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Disclaimer

signalword
Danger
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT RE 2
Storage Class
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service