Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34571

Supelco

2-(Diethyl-d10-amino)-6-methyl-4-pyrimidinol

PESTANAL®, analytical standard

Synonym(s):

2-(Diethyl-d10-amino)-6-hydroxy-4-methylpyrimidine, 2-(Diethyl-d10-amino)-6-methyl-1H-pyrimidin-4-one, DEAMPY-d10

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D10H5N3O
Molecular Weight:
191.30
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service