Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34565

Supelco

Monoquat iodide-(methyl-d3)

PESTANAL®, analytical standard

Synonym(s):

1-Methyl-d3-4,4′-bipyridinium iodide, 1-Methyl-d3-4-(4-pyridyl)pyridinium iodide, Monoquat-d3 iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11D3H8IN2
CAS Number:
Molecular Weight:
301.14
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

[2H]C([2H])([2H])[N+](C=C1)=CC=C1C2=CC=NC=C2.[I-]

General description

Monoquat iodide-(methyl-d3) is a deuterated analog of bipyridylium herbicide monoquat, a tertiary amine-quaternarized compound, wherein the methyl protons are substituted by deuterium.[1][2]

application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibacterial activity and characteristics of modified ferrite powder coated with a gemini pyridinium Salt molecule
SHIRAI A, et al.
Biocontrol Science, 12(3), 83-91 (2007)
Synthesis and properties of paraquat (methyl viologen) and other herbicidal alkyl homologs
Ross JH and Krieger RI
Journal of Agricultural and Food Chemistry, 28(5), 1026-1031 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service