Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34562

Supelco

Paraquat monopyridone iodide

PESTANAL®, analytical standard

Synonym(s):

1′,2′-Dihydro-1,1′-dimethyl-2′-oxo-4,4′-bipyridinium iodide, 1,1′-Dimethyl-2′-oxo-1′,2′-dihydro-[4,4′]bipyridinyl-1-ium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13IN2O
CAS Number:
Molecular Weight:
328.15
Beilstein/REAXYS Number:
4047961
UNSPSC Code:
12352202

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

C[N+](C=C1)=CC=C1C(C=CN2C)=CC2=O.[I-]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service