Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34536

Supelco

Pirimiphos-methyl-(O,O-dimethyl-d6)

PESTANAL®, analytical standard

Synonym(s):

2-Diethylamino-6-methylpyrimidin-4-yl-O,O-dimethyl-d6-phosphorothioate, Pirimiphos-methyl-d6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11D6H14N3O3PS
Molecular Weight:
311.37
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

CC1=NC(N(CC)CC)=NC(OP(OC([2H])([2H])[2H])(OC([2H])([2H])[2H])=S)=C1

InChI

1S/C11H20N3O3PS/c1-6-14(7-2)11-12-9(3)8-10(13-11)17-18(19,15-4)16-5/h8H,6-7H2,1-5H3/i4D3,5D3

InChI key

QHOQHJPRIBSPCY-RKAHMFOGSA-N

General description

Pirimiphos-methyl-(O,O-dimethyl-d6) is a deuterated analog of pirimiphos-methyl, an organophosphorus insecticide, wherein the O-methyl protons are substituted by deuterium.

Application

Pirimiphos-methyl-(O,O-dimethyl-d6) may be used as a deuterated internal standard to quantify pesticides in wheat flour and wastewater by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) and HPLC-MS/MS, respectively.
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of pesticides in wheat flour using microextraction on packed sorbent coupled to ultra-high performance liquid chromatography and tandem mass spectrometry
Di Ottavio F, et al.
Food Analytical Methods, 10(6), 1699-1708 (2017)
Evaluation of hazardous chemicals in edible insects and insect-based food intended for human consumption
Poma G, et al.
Food And Chemical Toxicology, 100, 70-79 (2017)
A robust analytical method for the determination of pesticide residues in wastewater
Jones L, et al.
Analytical Methods : Advancing Methods and Applications, 9(28), 4167-4174 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service