Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34515

Supelco

Phenmedipham-(methoxy-d3)

PESTANAL®, analytical standard

Synonym(s):

3-[(Methoxy-d3)carbonylamino]phenyl-N-(3-methylphenyl)carbamate, Phenmedipham-d3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D3H13N2O4
CAS Number:
Molecular Weight:
303.33
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

CC1=CC=CC(NC(OC2=CC=CC(NC(OC([2H])([2H])[2H])=O)=C2)=O)=C1

InChI

1S/C16H16N2O4/c1-11-5-3-6-12(9-11)18-16(20)22-14-8-4-7-13(10-14)17-15(19)21-2/h3-10H,1-2H3,(H,17,19)(H,18,20)/i2D3

InChI key

IDOWTHOLJBTAFI-BMSJAHLVSA-N

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service