Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34505

Supelco

Mecoprop-(4-chloro-2-methylphenoxy-d6)

PESTANAL®, analytical standard

Synonym(s):

2-(4-Chloro-2-methyl-d3-phenoxy-d3)propionic acid, Mecoprop-d6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D6H5ClO3
CAS Number:
Molecular Weight:
220.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24
Product 34505 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=C([2H])C([2H])=C(OC(C(O)=O)C)C(C([2H])([2H])[2H])=C1[2H]

InChI

1S/C10H11ClO3/c1-6-5-8(11)3-4-9(6)14-7(2)10(12)13/h3-5,7H,1-2H3,(H,12,13)/i1D3,3D,4D,5D

InChI key

WNTGYJSOUMFZEP-YFNZQJCKSA-N

General description

Mecoprop-(4-chloro-2-methylphenoxy-d6) is an isotope labeled analog of the post-emergence herbicide mecoprop, wherein the C-3, C-5, C-6 protons and C-2 methyl protons of the phenyl moiety are replaced by deuterium.[1]

application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis, potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mineralization of herbicide mecoprop by photoelectro-Fenton with UVA and solar light.
Flox C, et al.
Catalysis Today, 129(1-2), 29-36 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service