Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34504

Supelco

Trietazine-(monoethyl-d5-amino)

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-4-(diethylamino)-6-(ethyl-d5-amino)-1,3,5-triazine, 6-Chloro-N2-(ethyl-d5)-N4,N4-diethyl-1,3,5-triazine-2,4-diamine, Trietazine-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D5H11ClN5
CAS Number:
Molecular Weight:
234.74
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24
Product 34504 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=NC(N(CC)CC)=NC(NC([2H])([2H])C([2H])([2H])[2H])=N1

InChI

1S/C9H16ClN5/c1-4-11-8-12-7(10)13-9(14-8)15(5-2)6-3/h4-6H2,1-3H3,(H,11,12,13,14)/i1D3,4D2

InChI key

HFBWPRKWDIRYNX-SGEUAGPISA-N

General description

Trietazine-(monoethyl-d5-amino) is an isotope labeled analog of the pre-emergence herbicide trietazine, wherein the monoethyl protons are replaced by deuterium.[1]

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis, potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of herbicides on nodulation, symbiotic nitrogen fixation, growth and yield of pea (Pisum sativum)
Singh G and Wright D
Journal of Agricultural Science, 133(1), 21-30 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service