Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34483

Supelco

Propazine-(N2-isopropyl-d7)

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-4-(isopropyl-d7-amino)-6-(isopropylamino)-1,3,5-triazine, 6-Chloro-N2-(1-methylethyl-d7)-N4-(1-methylethyl)-1,3,5-triazine-2,4-diamine, Propazine-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D7H9ClN5
CAS Number:
Molecular Weight:
236.75
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)NC1=NC(Cl)=NC(NC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=N1

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service