34410
1,11-Dibromoundecane
purum, ≥98.0% (GC)
Synonym(s):
Undecamethylene dibromide
About This Item
Recommended Products
grade
purum
assay
≥98.0% (GC)
refractive index
n20/D 1.491 (lit.)
n20/D 1.492
bp
190-192 °C/18 mmHg (lit.)
mp
−10.6 °C (lit.)
density
1.335 g/mL at 25 °C (lit.)
SMILES string
BrCCCCCCCCCCCBr
InChI
1S/C11H22Br2/c12-10-8-6-4-2-1-3-5-7-9-11-13/h1-11H2
InChI key
SIBVHGAPHVRHMJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service