Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34377

Supelco

Trimethyltin chloride

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SnCl
CAS Number:
Molecular Weight:
199.27
Beilstein/REAXYS Number:
3535111
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

37-39 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

C[Sn](C)(C)Cl

InChI

1S/3CH3.ClH.Sn/h3*1H3;1H;/q;;;;+1/p-1

InChI key

KWTSZCJMWHGPOS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

206.6 °F - closed cup

flash_point_c

97 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangshu Zhai et al.
Chemosphere, 72(3), 389-399 (2008-04-25)
The photodegradation of methyltins, as environmental pollutants, has scarcely been studied so far because of the shortage of rapid and sensitive speciation methods, even though they have very simple structures. The photodegradation of monomethyltin trichloride (MMT), dimethyltin dichloride (DMT) and
Jiali Cai et al.
Chemical research in toxicology, 22(9), 1582-1587 (2009-08-07)
In evaluating the cytotoxic effects and the mechanisms of the apoptotic and necrotic actions of trimethyltin chloride (TMT) on human hepatoma G2 (HepG2) cells, the present study focused on the involvement of antiproliferation, DNA damage, cell death, apoptosis-related proteins, and
Erwin van Vliet et al.
Neurotoxicology, 28(6), 1136-1146 (2007-08-19)
Neurotoxicity aims to understand how xenobiotics interfere with the function of the nervous system and to unravel their mechanisms of action. Neuronal activity is the primary functional output of the nervous system and deviations from its resting level may indicate
Ya-ling Qian et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 26(8), 461-464 (2009-04-11)
To investigate suitable biomarkers for workers exposure to trimethyltin chloride (TMT-cl). Urinary samples of 44 male workers from five TMT-cl occupational poisoning incidents were collected. Methyltin mercaptide stabilizers and waste plastics used in the incidents were also collected. The levels
Na Zhao et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 29(4), 299-302 (2011-09-29)
To study the activity, protein and gene expression of renal HK-ATPase (HKA) in rats subchronic exposed to trimethyltin chloride (TMT). In subchronic toxic test (14-week), 55 female SD rats (age, 6 weeks) were divided randomly into 5 groups: control, low

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service