Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34316

Supelco

Calflo E

PESTANAL®, suitable for pesticide residue analysis

Synonym(s):

Calcium silicate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

product line

PESTANAL®

density

2.9 g/mL at 25 °C (lit.)

suitability

suitable for pesticide residue analysis

format

neat

SMILES string

[Ca++].[O-][Si]([O-])=O

InChI

1S/Ca.O3Si/c;1-4(2)3/q+2;-2

InChI key

OYACROKNLOSFPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Synthetic calcium silicate

Features and Benefits

Very low density, very high surface area

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongjie Hu et al.
Colloids and surfaces. B, Biointerfaces, 101, 83-90 (2012-07-17)
To improve the bioactivity and cytocompatibility of biomedical titanium dioxide coating, many efforts have been made to modify its surface composition and topography. Meanwhile, CaSiO(3) was commonly investigated as coating material on titanium implants for fast fixation and firm implant-bone
Li Yuan et al.
Langmuir : the ACS journal of surfaces and colloids, 29(2), 734-743 (2012-12-21)
A novel controlled drug-delivery system (CDDS) based on fluorescent mesoporous silica nanoparticles (FMSN) covalently linked with paclitaxel (PTX) via a disulfide linker was designed and characterized. A PTX prodrug based on a disulfide linker was synthesized, and its drug-delivery mechanism
Chen Wang et al.
Biomaterials, 34(1), 64-77 (2012-10-17)
As a potential bioactive material, β-calcium silicate (β-CS) has attracted particular attention in the field of bone regeneration. In this study, porous β-CS/Poly-D,L-Lactide-Glycolide (PDLGA) composite scaffolds were developed with the goals of controlling the degradation rate and improving the mechanical
X V Tran et al.
Journal of dental research, 91(12), 1166-1171 (2012-09-18)
In cases of pulp injury, capping materials are used to enhance tertiary dentin formation; Ca(OH)(2) and MTA are the current gold standards. The aim of this study was to evaluate the capacity of a new calcium-silicate-based restorative cement to induce
Aude Lacourt et al.
American journal of respiratory and critical care medicine, 187(9), 977-982 (2013-03-09)
Occupational coexposure to asbestos and other fibers or particles could modify the carcinogenicity of asbestos with regard to pleural mesothelioma. To estimate associations between pleural mesothelioma and occupational mineral wool and silica exposure and to study the impact of occupational

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service