Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

34280

1,3-Dibromopropane

purum, ≥99.0% (GC)

Synonym(s):

Trimethylene dibromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Br(CH2)3Br
CAS Number:
Molecular Weight:
201.89
EC Number:
203-690-3
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
635662
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

vapor density

7 (vs air)

grade

purum

assay

≥99.0% (GC)

refractive index

n20/D 1.524

bp

167 °C (lit.)

mp

−34 °C (lit.)

density

1.989 g/mL at 25 °C (lit.)

SMILES string

BrCCCBr

InChI

1S/C3H6Br2/c4-2-1-3-5/h1-3H2

InChI key

VEFLKXRACNJHOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

129.2 °F - closed cup

flash_point_c

54 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Witkowski et al.
The Journal of biological chemistry, 267(26), 18488-18492 (1992-09-15)
Thioesterase II is a 29-kDa monomer which, in certain specialized tissues, acts as a chain terminator in fatty acid synthesis by hydrolyzing medium-chain fatty acids from the fatty acid synthase. As with serine proteases, hydrolysis appears to involve acylation of
S P James et al.
Toxicology letters, 8(1-2), 7-15 (1981-04-01)
Oral administration of 1,3-dibromopropane (2 mmol/kg) to rats resulted in a marked decrease in the level of hepatic glutathione (GSH). Sulphur-containing [14C]-metabolites were excreted in the bile of rats dose with 1,3-bromo[14C]propane and were subjected to enterohepatic cycling. After an
E E Badr et al.
Journal of oleo science, 59(12), 647-652 (2010-11-26)
A fatty hydrazide based cationic gemini surfactants, 1,3-bis (N'-acyl-N,N-dimethylhydrazinium) propane dibromide which possess hydrolyzable amido moieties in the lipophilic portions, were prepared by reacting 1,3-bromopropane with N,N-dimethyl fatty hydrazide. The surface properties were explained and discussed based on the effect
Matthew S Nalewanski et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 61(7), 1547-1557 (2005-04-12)
The infrared and Raman spectrum of 1,3-dibromopropane is reported in the crystalline, liquid and gaseous states. These measurements are compared to the results of ab initio calculations carried out using the 6-31+g* Gaussian basis set for a restricted Hartree-Fock computation.
A R Jones et al.
Xenobiotica; the fate of foreign compounds in biological systems, 11(8), 541-546 (1981-08-01)
1. The metabolism of 1,3-dibromopropane had been investigated in the rat. Two conjugated metabolites have been isolated from the urine and identified as S-(3-hydroxypropyl)cysteine and N-acetyl-S-(3-hydroxypropyl)cysteine. 2. An oxidation product, identified as beta-bromolactic acid, has been isolated as a urinary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service