Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

34199

Supelco

PCB No 70

analytical standard

Synonym(s):

2,3′,4′,5-Tetrachlorobiphenyl, 2,3′,4′,5-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Cl4
CAS Number:
Molecular Weight:
291.99
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1ccc(Cl)c(c1)-c2ccc(Cl)c(Cl)c2

InChI

1S/C12H6Cl4/c13-8-2-4-10(14)9(6-8)7-1-3-11(15)12(16)5-7/h1-6H

InChI key

KENZYIHFBRWMOD-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Health hazardEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service