Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

34188

Supelco

TBPP

PESTANAL®, analytical standard

Synonym(s):

Tris(2,3-dibromopropyl) phosphate, TDBPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15Br6O4P
CAS Number:
Molecular Weight:
697.61
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

BrCC(Br)COP(=O)(OCC(Br)CBr)OCC(Br)CBr

InChI

1S/C9H15Br6O4P/c10-1-7(13)4-17-20(16,18-5-8(14)2-11)19-6-9(15)3-12/h7-9H,1-6H2

InChI key

PQYJRMFWJJONBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 1B - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M W Jernaes et al.
Cytometry, 17(4), 302-309 (1994-12-01)
In an attempt to develop procedures for nucleic acid staining of bacteria for clinical routine assays, the uptake of ethidium bromide (EB) in wild-type Escherichia coli was studied using flow cytometry. Phosphate-buffered saline (PBS) containing EDTA or Tris significantly increased
Nadeem Ali et al.
Chemosphere, 88(11), 1276-1282 (2012-05-04)
Due to worldwide restrictions on polybrominated diphenyl ethers (PBDEs), the demand for alternative flame retardants (AFRs), such as organophosphate flame retardants (OPFRs), novel brominated FRs (NBFRs) and hexabromocyclododecanes (HBCDs), has recently increased. Little is known about human exposure to NBFRs
Phil Brown et al.
Health affairs (Project Hope), 30(5), 906-914 (2011-05-11)
Flame retardant chemicals, used in many furniture and electronic products to slow combustion, are an example of science and policy problems related to chemical usage. Drawing on a literature review and interviews with stakeholders, we argue that early warnings of
G J Van Beerendonk et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 28(1), 111-117 (1995-11-01)
The metabolism of tris(2,3-dibromopropyl) phosphate (Tris-BP) was compared with that of completely deuterated Tris-BP (D15-Tris-BP) in an isolated, recirculating rat liver perfusion system in order to determine the relative quantitative importance of two different biotransformation pathways of Tris-BP: (i) cytochrome
Katherine M Morton et al.
Reproduction, fertility, and development, 19(7), 792-796 (2007-09-28)
Epididymal spermatozoa were harvested from male alpacas and frozen after extension and cooling to 4 degrees C in citrate-, Tris- and lactose-based diluents (Experiment 1) and as pellets in 0.25- and 0.5-mL straws on either dry ice or over liquid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service