Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34129

Supelco

3-Acetyl-d3-deoxynivalenol solution

100 μg/mL in acetonitrile, analytical standard

Synonym(s):

15-Acetylvomitoxin, 3-d3-AcDON

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17D3H19O7
CAS Number:
Molecular Weight:
341.37
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

mass shift

M+3

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])C(=O)O[C@@H]1C[C@@]2(C)[C@]3(CO3)[C@@H]1O[C@@H]4C=C(C)C(=O)[C@@H](O)[C@]24CO

InChI

1S/C17H22O7/c1-8-4-11-16(6-18,13(21)12(8)20)15(3)5-10(23-9(2)19)14(24-11)17(15)7-22-17/h4,10-11,13-14,18,21H,5-7H2,1-3H3/t10-,11-,13-,14-,15-,16-,17+/m1/s1/i2D3

InChI key

ADFIQZBYNGPCGY-POMOKOCZSA-N

General description

3-Acetyl-d3-deoxynivalenol is an isotopically labeled standard of the trichothecene mycotoxin, 3-acetyl-deoxynivalenol (3-Ac-DON), mainly produced by molds of the genus, Fusarium.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F

flash_point_c

2.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of mycotoxins in bovine milk by liquid chromatography tandem mass spectrometry.
Sorensen KL and Elbaek HT
Journal of Chromatography. B, Biomedical Sciences and Applications, 820(2), 183-196 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service