34120
BDE No 209 solution
50 μg/mL in isooctane:toluene (9:1), analytical standard
Synonym(s):
Decabromodiphenyl ether solution, Decabromodiphenyl oxide
About This Item
Recommended Products
grade
analytical standard
shelf life
limited shelf life, expiry date on the label
concentration
50 μg/mL in isooctane:toluene (9:1)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
single component solution
storage temp.
2-8°C
SMILES string
Brc1c(Br)c(Br)c(Oc2c(Br)c(Br)c(Br)c(Br)c2Br)c(Br)c1Br
InChI
1S/C12Br10O/c13-1-3(15)7(19)11(8(20)4(1)16)23-12-9(21)5(17)2(14)6(18)10(12)22
InChI key
WHHGLZMJPXIBIX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
application
- House dust samples by liquid chromatography with negative-ion atmospheric pressure photoionization tandem mass spectrometry (LC/NI-APPI/MS/MS).
- Sediment samples by selective pressurized liquid extraction (SPLE) followed by analysis using gas chromatography (GC) combined with negative chemical ionization (NCI)−MS in selective ion monitoring (SIM) mode.
- Sewage sludge by GC-SIM with low resolution MS and in blood, muscle as well as liver samples of European starlings (Sturnus vulgaris) by GC-MS.
signalword
Danger
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
10.4 °F - closed cup
flash_point_c
-12 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service