Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34026

Supelco

(±)-4,4,4-Trifluoro-3-(3-indolyl)butyric acid

PESTANAL®, analytical standard

Synonym(s):

(±)-β-(Trifluoromethyl)-3-indolepropionic acid, TFIBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10F3NO2
CAS Number:
Molecular Weight:
257.21
UNSPSC Code:
41116107

grade

analytical standard

product line

PESTANAL®

storage temp.

2-8°C

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service