Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33986

Supelco

Tiopronin

VETRANAL®, analytical standard

Synonym(s):

N-(2-Mercaptopropionyl)glycine, Tiopronin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(SH)CONHCH2COOH
CAS Number:
Molecular Weight:
163.19
Beilstein/REAXYS Number:
1859822
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(S)C(=O)NCC(O)=O

InChI

1S/C5H9NO3S/c1-3(10)5(9)6-2-4(7)8/h3,10H,2H2,1H3,(H,6,9)(H,7,8)

InChI key

YTGJWQPHMWSCST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mildred Quintana et al.
Chemical communications (Cambridge, England), 48(100), 12159-12161 (2012-10-24)
Under ultrasonication, the production of high quality graphene layers by exfoliation of graphite was achieved via addition of tiopronin as an antioxidant.
Self-assembled ultra-high aspect ratio silver nanochains.
Matthew J Shiers et al.
Advanced materials (Deerfield Beach, Fla.), 24(38), 5227-5235 (2012-08-07)
Andrew S Goldsborough et al.
Journal of medicinal chemistry, 54(14), 4987-4997 (2011-06-11)
A major challenge in the treatment of cancer is multidrug resistance (MDR) that develops during chemotherapy. Here we demonstrate that tiopronin (1), a thiol-substituted N-propanoylglycine derivative, was selectively toxic to a series of cell lines expressing the drug efflux pump
Xiaoqing Cai et al.
Analytical and bioanalytical chemistry, 401(3), 809-816 (2011-05-04)
Well-dispersed gold nanoparticles (NP) coated with tiopronin were synthesized by X-ray irradiation without reducing agents. High-resolution transmission electron microscopy shows that the average core diameters of the NPs can be systematically controlled by adjusting the tiopronin to Au mole ratio
Jung Ah Lee et al.
Journal of Korean medical science, 27(5), 547-552 (2012-05-09)
We previously demonstrated that there are acute and delayed phases of renal protection against renal ischemia and reperfusion (IR) injury with renal ischemic preconditioning (IPC). This study assessed whether hepatic IPC could also reduce distant renal IR injury through the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service