Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33978

Supelco

2-Acetamido-5-nitrothiazole

VETRANAL®, analytical standard

Synonym(s):

N-(5-Nitro-2-thiazolyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N3O3S
CAS Number:
Molecular Weight:
187.18
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(=O)Nc1ncc(s1)[N+]([O-])=O

InChI

1S/C5H5N3O3S/c1-3(9)7-5-6-2-4(12-5)8(10)11/h2H,1H3,(H,6,7,9)

InChI key

UJRRDDHEMZLWFI-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Search for contact spermatocidal agents].
L N Kopteva
Akusherstvo i ginekologiia, (2)(2), 67-70 (1992-02-01)
I I Oleĭnik et al.
Stomatologiia, (6)(6), 8-10 (1991-11-01)
Twenty-three patients with generalized periodontitis were examined. Microbiologic examination of the periodontal pouch contents, making use of anaerobic cultivation, resulted in isolation and identification of 103 strains, 76 (72.8%) of which were referred to obligate anaerobes. Facultative anaerobes made up
N F Kalinichenko
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 60(1), 83-91 (1998-06-11)
The antibacterial activity of protistocide drug nitazole has been revealed for the first time in the 80's at the Laboratory of Clinical Microbiology of the Mechnikov Research Institute of Microbiology and Immunology. Unlike other imidazoles, such as metronidazole and tinidazole
S V Tarasenko et al.
Stomatologiia, 79(2), 17-19 (2000-05-17)
Combined drugs with a hydrophilic base, nitacide and hyposol-n, were used in combined therapy of acute purulent inflammations of the maxillofacial area. Time course of clinical parameters and changes in microflora were studied in patients administered different local treatments with
J P Raynaud et al.
American journal of veterinary research, 42(1), 51-53 (1981-01-01)
A swine dysentery (SD) model that produces consistent, homogeneous, and severe SD was used in 2 experiments to compare the prophylactic effectiveness of 5 commercially available swine feed additive products. Under the conditions of these studies, carbadox and carbadox +

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service