Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33967

Supelco

Sparfloxacin

VETRANAL®, analytical standard

Synonym(s):

5-Amino-1-cyclohexyl-7-(cis-3,5-dimethylpiperazino)-6,8-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22F2N4O3
CAS Number:
Molecular Weight:
392.40
Beilstein/REAXYS Number:
9170271
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

C[C@H]1CN(C[C@@H](C)N1)c2c(F)c(N)c3C(=O)C(=CN(C4CC4)c3c2F)C(O)=O

InChI

1S/C19H22F2N4O3/c1-8-5-24(6-9(2)23-8)17-13(20)15(22)12-16(14(17)21)25(10-3-4-10)7-11(18(12)26)19(27)28/h7-10,23H,3-6,22H2,1-2H3,(H,27,28)/t8-,9+

InChI key

DZZWHBIBMUVIIW-DTORHVGOSA-N

Looking for similar products? Visit Product Comparison Guide

application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aura Rusu et al.
Journal of chromatographic science, 52(8), 919-925 (2013-08-03)
The migration behavior and separation of 13 quinolone antibacterials were investigated by capillary electrophoresis (CE). In order to predict the electrophoretic mobility, the protonation macroconstants of all the compounds were determined by pH-potentiometric titrations. We proved that the electrophoretic mobility
Artur Beberok et al.
Pharmacological reports : PR, 67(1), 38-43 (2015-01-07)
Fluoroquinolones are a group of broad spectrum bactericidal antibiotics used to treat various infections of urinary and respiratory systems, as well as in ophthalmology and dermatology. This class of antibiotics causes toxic effects directed to pigmented tissues, what introduces a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service