Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33947

Supelco

T-2 Toxin

reference material

Synonym(s):

12,13-Epoxytrichothec-9-ene-3,4,8,15-tetrol-4,15-diacetate-8-isovalerate, 4β,15-Diacetoxy-3α-hydroxy-8α-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34O9
CAS Number:
Molecular Weight:
466.52
Beilstein/REAXYS Number:
3575695
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
reference material

shelf life

limited shelf life, expiry date on the label

availability

not available in USA

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

Storage temp.

−20°C

SMILES string

[H][C@]12O[C@]3([H])[C@H](O)[C@@H](OC(C)=O)[C@](C)([C@@]1(COC(C)=O)C[C@H](OC(=O)CC(C)C)C(C)=C2)[C@]34CO4

InChI

1S/C24H34O9/c1-12(2)7-18(27)32-16-9-23(10-29-14(4)25)17(8-13(16)3)33-21-19(28)20(31-15(5)26)22(23,6)24(21)11-30-24/h8,12,16-17,19-21,28H,7,9-11H2,1-6H3/t16-,17+,19+,20+,21+,22+,23+,24-/m0/s1

Inchi Key

BXFOFFBJRFZBQZ-QYWOHJEZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

T-2 Toxin is found to be a potent trichothecene mycotoxin produced by the genus fusarium fungi in feedstuffs and cereal grains.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Skin Irrit. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of dietary nucleotides in reduction of DNA damage induced by T-2 toxin and deoxynivalenol in chicken leukocytes
Frankie.T, et al.
Food And Chemical Toxicology, 44, 1838-1844 (2006)
Yanshen Li et al.
Journal of agricultural and food chemistry, 59(8), 3441-3453 (2011-03-23)
This review focuses on the toxicity and metabolism of T-2 toxin and analytical methods used for the determination of T-2 toxin. Among the naturally occurring trichothecenes in food and feed, T-2 toxin is a cytotoxic fungal secondary metabolite produced by
Soujanya Ratna Edupuganti et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 923-924, 98-101 (2013-03-19)
An affinity purification method that isolates T-2 toxin-specific IgY utilizing a T-2-toxin-immobilized column was developed. The T-2 toxin was covalently coupled via a carbonyldiimidazole-activated hydroxyl functional group to amine-activated sepharose beads. The affinity-purified IgY was characterized by gel electrophoresis, fast
A Osselaere et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 55, 150-155 (2013-01-15)
The effects of the mycotoxin T-2 on hepatic and intestinal drug-metabolizing enzymes (cytochrome P450) and drug transporter systems (MDR1 and MRP2) in poultry were investigated during this study. Broiler chickens received either uncontaminated feed, feed contaminated with 68μg/kg or 752μg/kg
Elin Verbrugghe et al.
Research in veterinary science, 93(3), 1139-1141 (2012-07-28)
The aim of the study was to investigate the effect of a modified glucomannan binder on the course of a Salmonella Typhimurium infection in pigs. Therefore, four pig diets were provided during 23 days: (1) free of mycotoxins, (2) containing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service