Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33865

Supelco

DTPD

analytical standard

Synonym(s):

N,N′-Di-(p-tolyl)-p-phenylenediamine, N,N′-(p-Phenylene)ditoluidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20N2
CAS Number:
Molecular Weight:
288.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

CC1=CC=C(NC2=CC=C(NC3=CC=C(C)C=C3)C=C2)C=C1

InChI

1S/C20H20N2/c1-15-3-7-17(8-4-15)21-19-11-13-20(14-12-19)22-18-9-5-16(2)6-10-18/h3-14,21-22H,1-2H3

InChI key

JBMMSVXRQPXCGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Standard for the assay of additives in rubber[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Wolfe et al.
The Journal of organic chemistry, 65(4), 1158-1174 (2000-05-18)
Palladium complexes supported by (o-biphenyl)P(t-Bu)(2) (3) or (o-biphenyl)PCy(2) (4) are efficient catalysts for the catalytic amination of a wide variety of aryl halides and triflates. Use of ligand 3 allows for the room-temperature catalytic amination of many aryl chloride, bromide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service