Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33863

Supelco

Bensultap

PESTANAL®, analytical standard

Synonym(s):

S,S′-[2-(Dimethylamino)trimethylene] di(benzenethiosulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO4S4
CAS Number:
Molecular Weight:
431.61
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CN(C)C(CSS(=O)(=O)c1ccccc1)CSS(=O)(=O)c2ccccc2

InChI

1S/C17H21NO4S4/c1-18(2)15(13-23-25(19,20)16-9-5-3-6-10-16)14-24-26(21,22)17-11-7-4-8-12-17/h3-12,15H,13-14H2,1-2H3

InChI key

YFXPPSKYMBTNAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bensultap is a broad spectrum, carbamate insecticide used for the control of pests such as leaf rollers, beetles, leaf miners, borers, boll, root weevils, and various moths in crop and fruit plantations. Its mode of action involves the blocking of synaptic neuron transmissions in the central nervous system (CNS) of insect pests.[1]

Application

Bensultap may be used as an analytical reference standard for the determination of the analyte in crop and soil samples using high performance liquid chromatography and gas chromatography with flame photometric detection.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Pesticide Encyclopedia (2014)
Viktor Szegedi et al.
Toxicology, 214(1-2), 67-76 (2005-07-13)
In the present multilevel study, neuromodulatory effect of two insecticides, bensultap and fipronil were investigated in rats. Although the new generation of insecticides shows greater affinity to invertebrate as compared to mammalian receptors, toxic effect of these compounds in vertebrates
A M Grigor'ev et al.
Sudebno-meditsinskaia ekspertiza, 52(5), 30-35 (2010-01-12)
This paper is designed to describe methods for the detection of derivatives and metabolites of the insecticidic agent bancol (nereistoxin precursor) by chromatographic methods including thin layer chromatography (TLC), high-performance liquid chromatography (HPLC), gas chromatography with a nitrogen-phosphorous detector (GC-NPD)
Toxicity of three insecticides in a standard algal growth inhibition test with Scenedesmus subspicatus.
K Burkiewicz et al.
Bulletin of environmental contamination and toxicology, 74(6), 1192-1198 (2005-09-15)
János Györi et al.
Toxicology in vitro : an international journal published in association with BIBRA, 21(6), 1050-1057 (2007-05-18)
Electrophysiological experiments were performed on in vitro neuronal preparations from terrestrial snail and rat brain slices, to determine the effect of the insecticide bensultap. Although bensultap has low toxicity in mammals, our results showed that bensultap altered the synaptic transmission

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service