Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33854

Supelco

Thionazin solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Synonym(s):

O,O-Diethyl O-(2-pyrazinyl) phosphorothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13N2O3PS
CAS Number:
Molecular Weight:
248.24
Beilstein/REAXYS Number:
884916
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1cnccn1

InChI

1S/C8H13N2O3PS/c1-3-11-14(15,12-4-2)13-8-7-9-5-6-10-8/h5-7H,3-4H2,1-2H3

InChI key

IRVDMKJLOCGUBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thionazin is a pre- and post-emergence,[1] organophosphorus pesticide (OPP), extensively used in agriculture to combat agricultural pests.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Thionazin may be used as an analytical reference standard for the determination of the analyte in :
  • Soil and marine sediment samples using supercritical fluid extraction with dispersive liquid–liquid microextraction followed by gas chromatography-flame ionization detection (GC-FID).[2]
  • Human blood samples using liquid-liquid extraction followed by gas chromatography and liquid chromatography coupled to electrospray ionization-tandem mass spectrometry in multiple reaction monitoring (MRM) mode.[3]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F

flash_point_c

2 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combination of supercritical fluid extraction with dispersive liquid-liquid microextraction for extraction of organophosphorus pesticides from soil and marine sediment samples.
Naeeni HM, et al.
Journal of Supercritical Fluids, 57(3), 219-226 (2011)
Soil disinfestation.
Developments in Agricultural and Managed-Forest Ecology, 6(5), 307-315 (2012)
Validated analytical methodology for the simultaneous determination of a wide range of pesticides in human blood using GC--MS/MS and LC--ESI/MS/MS and its application in two poisoning cases.
Luzardo PO, et al.
Science & Justice, 55(5), 307-315 (2015)
Maya Kochman et al.
Journal of chromatography. A, 1129(1), 95-104 (2006-07-13)
A new approach of flow modulation comprehensive two-dimensional gas chromatography-mass spectrometry (GC x GC-MS) with supersonic molecular beam (SMB) and a quadrupole mass analyzer is presented. Flow modulation uniquely enables GC x GC-MS to be achieved even with the limited
F Sepulcri et al.
Schweizerische medizinische Wochenschrift, 117(13), 487-490 (1987-03-28)
We report on a case of severe intoxication in an 18-year-old female who had ingested an insecticide with suicidal intent. The insecticide (thionacine) is a cholinesterase inhibitor used in gardening and agriculture. The resources of modern intensive care permitted successful

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service