33709
Tetrahydrofuran
≥99.9% (GC), contains ~250 mg/L 2, 6-di-tert.-butyl-4-methylphenol as stabilizer, puriss. p.a.
Synonym(s):
THF, Butylene oxide, Oxolane, Tetramethylene oxide
About This Item
143 mmHg ( 20 °C)
Recommended Products
Product Name
Tetrahydrofuran, puriss. p.a., ≥99.9% (GC)
vapor density
2.5 (vs air)
vapor pressure
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)
grade
puriss. p.a.
assay
≥99.9% (GC)
form
liquid
autoignition temp.
610 °F
contains
~250 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer
expl. lim.
1.8-11.8 %
technique(s)
GC/GC: suitable
impurities
≤0.001% free acid (as CH3COOH)
≤0.001% non-volatile matter
≤0.005% peroxides (as H2O2)
≤0.05% water (Karl Fischer)
color
APHA: <20
refractive index
n20/D 1.407 (lit.)
bp
65-67 °C (lit.)
mp
−108 °C (lit.)
solubility
H2O: soluble
density
0.889 g/mL at 25 °C (lit.)
cation traces
Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg
SMILES string
C1CCOC1
InChI
1S/C4H8O/c1-2-4-5-3-1/h1-4H2
InChI key
WYURNTSHIVDZCO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
Go to our Greener Alternative-Cyclopentyl methylether
A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
recommended
suggested gloves for splash protection
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system, Respiratory system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
-6.2 °F - closed cup
flash_point_c
-21.2 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service