Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33707

Supelco

4-(3,5-Dimethyl-3-heptyl)phenol

analytical standard

Synonym(s):

353 Nonylphenol, 4-(1-Ethyl-1,3-dimethylpentyl)phenol, p-353-NP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
Beilstein/REAXYS Number:
9325718
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

−20°C

SMILES string

CCC(C)CC(C)(CC)c1ccc(O)cc1

InChI

1S/C15H24O/c1-5-12(3)11-15(4,6-2)13-7-9-14(16)10-8-13/h7-10,12,16H,5-6,11H2,1-4H3

InChI key

RYIHVIPUIXFRNI-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service