Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33693

Supelco

Equilenin solution

VETRANAL®, 100 μg/mL in acetonitrile, analytical standard

Synonym(s):

3-Hydroxy-1,3,5(10),6,8-estrapentaen-17-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O2
CAS Number:
Molecular Weight:
266.33
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC(=O)[C@@]1(C)CCc3c2ccc4cc(O)ccc34

InChI

1S/C18H18O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h2-5,10,16,19H,6-9H2,1H3/t16-,18-/m0/s1

InChI key

PDRGHUMCVRDZLQ-WMZOPIPTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Equilenin may be used as an analytical reference standard for the quantification of the analyte in pharmaceutical formulations using high-performance liquid chromatography (HPLC) with fluorescence and ultraviolet detection.[1][2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F

flash_point_c

2 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William Childs et al.
Journal of the American Chemical Society, 132(18), 6474-6480 (2010-04-20)
A light-activated reaction analog has been developed to mimic the catalytic reaction cycle of Delta(5)-3-ketosteroid isomerase to probe the functionally relevant protein solvation response to the catalytic charge transfer. Delta(5)-3-ketosteroid isomerase from Pseudomonas putida catalyzes a C-H bond cleavage and
Minsun Chang et al.
Advances in experimental medicine and biology, 617, 601-607 (2008-05-24)
Oxidative metabolism of estrogens has been associated with genotoxicity. O-methylation of catechol estrogens is considered as a protective mechanism. 4-Methoxyequilenin (4-MeOEN) is the O-methylated product of 4-hydroxyequilenin (4-OHEN). 4-OHEN, the major catechol metabolite of the equine estrogens present in the
Shuang Ding et al.
Chemical research in toxicology, 21(5), 1064-1073 (2008-04-18)
Equilin and equilenin, components of the hormone replacement therapy drug Premarin, can be metabolized to the catechol 4-hydroxyequilenin (4-OHEN). The quinoids produced by 4-OHEN oxidation react with dC, dA, and dG to form unusual stable cyclic adducts, which have been
The Pesticide Encyclopedia, 17(2), 337-347 (2014)
Kuan-Wei Peng et al.
Chemical research in toxicology, 23(8), 1374-1383 (2010-06-15)
4-Hydroxyequilenin (4-OHEN) is a major phase I metabolite of the equine estrogens present in widely prescribed hormone replacement formulations. 4-OHEN is autoxidized to an electrophilic o-quinone that has been shown to redox cycle, generating ROS, and to covalently modify proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service