Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33671

Supelco

BDE No 49 solution

50 μg/mL in isooctane, analytical standard

Synonym(s):

2,2′,4,5′-Tetrabromodiphenyl ether solution, 2,2′,4,5′-TetraBDE, PBDE 49

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Br4O
CAS Number:
Molecular Weight:
485.79
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

50 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Brc1ccc(Oc2cc(Br)ccc2Br)c(Br)c1

InChI

1S/C12H6Br4O/c13-7-2-4-11(10(16)5-7)17-12-6-8(14)1-3-9(12)15/h1-6H

InChI key

QWVDUBDYUPHNHY-UHFFFAOYSA-N

General description

BDE No 49 is a ubiquitous environmental contaminant belonging to the class of polybrominated diphenyl ethers.[1] It can find wide applications as a flame-retardants additive in consumer products ranging from textiles to polymers.

Application

BDE No 49 may be used as an analytical reference standard for the determination of the analyte in milk and dust samples using gas chromatography with mass spectrometric detection (GC-MS).[2][3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

The customer needs to make sure that the standard is homogeneous, this can be achieved by shaking, vortexing , or sonicating the ampoule before opening the ampoule. Care should be taken when opening the ampoule to avoid any injuries from the broken glass.
The standard should be transferred to a clean and appropriate vial or flask using clean pipettes or micro pipettes. The vial should be immediately capped to avoid any loss or evaporation of the solvent.
After opening the ampoule, the standard should not be stored or kept in the ampoule. To preserve the integrity of the product, the standard should be transferred to an appropriate vial that must be capped and stored according to the recommendation on the Certificate of Analysis.

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

10.4 °F

flash_point_c

-12 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of polybrominated diphenyl ethers in human milk samples in the Czech Republic: comparative study of negative chemical ionisation mass spectrometry and time-of-flight high-resolution mass spectrometry.
Kazda R, et al.
Analytica Chimica Acta, 520(1-2), 237-243 (2004)
Clean-up method for determination of established and emerging brominated flame retardants in dust.
Sahlstrom L, et al.
Analytical and Bioanalytical Chemistry, 404(2), 459-466 (2012)
GC / MS analysis of polybrominated diphenyl ethers in fish collected from the Inland Sea of ??Seto, Japan.
Akutsu K, et al.
Chemosphere, 44(6), 1325-1333 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service