Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33572

Supelco

4-(3,6-Dimethyl-3-heptyl)phenol monoethoxylate-ring-13C6 solution

10 μg/mL in acetone, analytical standard

Synonym(s):

2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy]ethanol-13C6, 2-[4-(3,6-Dimethyl-3-heptyl)phenoxy]ethanol-13C6, 3,6,3-Nonylphenol-monoethoxylate-13C6, 363-NP1EO-13C6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C11H28O2
CAS Number:
Molecular Weight:
270.36
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

mass shift

M+6

storage temp.

−20°C

SMILES string

CCC(C)(CCC(C)C)[13c]1[13cH][13cH][13c](OCCO)[13cH][13cH]1

InChI

1S/C17H28O2/c1-5-17(4,11-10-14(2)3)15-6-8-16(9-7-15)19-13-12-18/h6-9,14,18H,5,10-13H2,1-4H3/i6+1,7+1,8+1,9+1,15+1,16+1

InChI key

QRFCMJANHDRHEH-CJFYFUKZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

Application

4-(3,6-Dimethyl-3-heptyl)phenol monoethoxylate-ring-13C6 solution is an analytical labeld standard used for the determination of mono and di-ethoxylates of nonylphenol (NP) and octylphenol (OP) in environmental samples using solid-phase micro extraction followed by gas chromatography with mass spectrometry (SPME-GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

1.4 °F - closed cup

flash_point_c

-17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Alkylphenols are used in various industries like paper manufacturing and pesticide formulations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service