Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33571

Supelco

4-(3,6-Dimethyl-3-heptyl)phenol-3,5-d2 solution

1 μg/mL in acetone, analytical standard

Synonym(s):

3,6,3-Nonylphenol-d2, 363-NP-D2, 4-(1-Ethyl-1,4-dimethylpentyl)phenol-3,5-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D2H22O
CAS Number:
Molecular Weight:
222.36
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

1 μg/mL in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

mass shift

M+2

storage temp.

−20°C

SMILES string

[2H]c1cc(O)cc([2H])c1C(C)(CC)CCC(C)C

InChI

1S/C15H24O/c1-5-15(4,11-10-12(2)3)13-6-8-14(16)9-7-13/h6-9,12,16H,5,10-11H2,1-4H3/i6D,7D

InChI key

BGLUSMSXKMBLFS-QFIQSOQBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

1.4 °F - Information refers to the main ingredient.

flash_point_c

-17 °C - Information refers to the main ingredient.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Alkylphenols are used in various industries like paper manufacturing and pesticide formulations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service