Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33402

Supelco

TNP Mix 11 solution

1 mL in methanol, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

1 mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Components

45316 Alachlor 100ng/μl +/- 0,5
45370 Carbofuran 100ng/μl +/- 0,5
45431 Dichlorbenzil 100ng/μl +/- 0,5
45566 Metamitron 100ng/μl +/- 0,5
45567 Metazachlor 100ng/μl +/- 0,5
45579 Metolachlor 100ng/μl +/- 0,5
45581 Metribuzin 100ng/μl +/- 0,5
45612 Pendimethalin 100ng/μl +/- 0,5
45677 Terbutryn 100ng/μl +/- 0,5
45695 Triallat 100ng/μl +/- 0,5
45700 Ttrifluralin 100ng/μl +/- 0,5

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service