Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33367

Supelco

Methyl-pentachlorophenylsulfide

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H3Cl5S
CAS Number:
Molecular Weight:
296.43
Beilstein/REAXYS Number:
2214412
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

92-96 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

CSc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C7H3Cl5S/c1-13-7-5(11)3(9)2(8)4(10)6(7)12/h1H3

InChI key

LGZZJTIUEJNNKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Bakke et al.
Xenobiotica; the fate of foreign compounds in biological systems, 20(6), 601-605 (1990-06-01)
1. Biliary metabolites from rats dosed with pentachlorothioanisole (PCTA) were characterized by fast atom bombardment mass spectrometry and electron impact mass spectrometry. 2. Most of the biliary metabolites from PCTA were mercapturic acid pathway metabolites of methylsulphinyltetrachlorobenzene (51% of the
Artifact formation during derivatization of methylsulfinyl-containing metabolites for electron impact mass spectrometry.
J E Bakke
Biomedical & environmental mass spectrometry, 19(10), 635-635 (1990-10-01)
Y Kato et al.
Journal of pharmacobio-dynamics, 13(5), 278-284 (1990-05-01)
Effects of hexachlorobenzene (HCB) and its sulfur-containing metabolites on the heme metabolic enzymes in rat liver were investigated. A single injection of HCB caused the increase in activities of delta-aminolevulinic acid (ALA) synthetase and heme oxygenase, and contents of cytochrome
D J Mulford et al.
Xenobiotica; the fate of foreign compounds in biological systems, 21(5), 597-603 (1991-05-01)
1. The metabolism of pentachlorothioanisole to bis-(methylthio)tetrachlorobenzene was shown to involve turnover of about 50% of the original methylthio group. 2. Metabolites of the displaced methylthio group were methanesulphinic acid and sulphate in the urine, carbon dioxide in the expired
Hexachlorobenzene (HCB) and some of its sulfur-containing metabolites: tissue retention and excretion routes in mice.
I Brandt et al.
Developments in toxicology and environmental science, 8, 627-630 (1980-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service