Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33340

Sigma-Aldrich

4,5-Diaminopyrimidine

purum, ≥95.0% (NT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4
CAS Number:
Molecular Weight:
110.12
Beilstein/REAXYS Number:
112646
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Quality Level

assay

≥95.0% (NT)

bp

229 °C/32 mmHg (lit.)

mp

~200 °C
204-206 °C (lit.)

SMILES string

Nc1cncnc1N

InChI

1S/C4H6N4/c5-3-1-7-2-8-4(3)6/h1-2H,5H2,(H2,6,7,8)

InChI key

PPAULTVPKLVLII-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may discolor to yellow on storage

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service